Product Name
DEET PESTANAL®, analytical standard
Product Number
36542
Product Brand
SIAL
CAS Number
134-62-3
Molecular Weight
191.27
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow to Brown-Yellow
APPEARANCE (FORM)Liquid
PURITY (HPLC AREA %)≥ 95.0 %
REFRACTIVE INDEX N20/D1.5200 - 1.5240
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS