Product Name
Cumene PESTANAL®
Product Number
36698
Product Brand
SIAL
CAS Number
98-82-8
Molecular Weight
120.19
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.4890 - 1.4930
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS