Product Name
1,2-Dichlorobenzene PESTANAL®, analytical standard
Product Number
36707
Product Brand
SIAL
CAS Number
95-50-1
Molecular Weight
147.00
Test
Specification
APPEARANCE (COLOR)Colorless to Almost Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
DENSITY D20/41.280 - 1.320
REFRACTIVE INDEX N20/D1.5496 - 1.5536
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS