Product Name
Chlorfenapyr PESTANAL®, analytical standard
Product Number
37913
Product Brand
SIAL
CAS Number
122453-73-0
Molecular Weight
407.61
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT99 - 104 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS