Product Name
Simvastatin analytical standard
Product Number
38956
Product Brand
SIAL
CAS Number
79902-63-9
Molecular Weight
418.57
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 98.0 %
SPECIFIC ROTATION (20/D)275.0 ± 25.0 Degrees
CONCENTRATIONC=1 in ACETONITRILE
WATER≤ 0.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS