Product Name
3,3′-Diethylthiacyanine iodide Dye content ~97 %
Product Number
390410
Product Brand
SIAL
CAS Number
2197-01-5
Molecular Weight
466.40
Test
Specification
Appearance (Color)Conforms to Requirements
 Yellow to Dark Green
Appearance (Form)Conforms to Requirement
 Powder or Crystals
Infrared spectrumConforms to Structure
Wavelength420 - 428 nm
 0.004g/l, Ethanol
Extinction Coefficient≥65000
Carbon47.2 - 50.6 %
Nitrogen5.7 - 6.2 %
Purity (HPLC)≥96.5 %