Product Name
2-Methylfuran analytical standard
Product Number
39587
Product Brand
SIAL
CAS Number
534-22-5
Molecular Weight
82.10
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow and Colorless to Green-Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.0 %
REFRACTIVE INDEX N20/D1.432 - 1.434
WATER (COULOMETR.)≤ 0.3 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS