Product Name
Carvacrol analytical standard
Product Number
42632
Product Brand
SIAL
CAS Number
499-75-2
Molecular Weight
150.22
Test
Specification
APPEARANCE (COLOR)Colorless to Brown-Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.523 - 1.525
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS