Product Name
Cyclohexene analytical standard
Product Number
44028
Product Brand
SIAL
CAS Number
110-83-8
Molecular Weight
82.14
Test
Specification
STABILIZERSTABILIZED WITH ~0.01 % 2,6-DI-TERT-BUTYL-4-METHYLPHENOL
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.7 %
REFRACTIVE INDEX N20/D1.445 - 1.447
WATER (COULOMETR.)≤ 0.05 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS