Product Name
Fluridon PESTANAL®, analytical standard
Product Number
45511
Product Brand
SIAL
CAS Number
59756-60-4
Molecular Weight
329.32
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow to Faint Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT151 - 157 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS