Product Name
Methyl isothiocyanate PESTANAL®, analytical standard
Product Number
45576
Product Brand
SIAL
CAS Number
556-61-6
Molecular Weight
73.12
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals or Solid or Chunks
PURITY (GC AREA %)≥ 98.0 %
MELTING POINT34 - 39 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS