Product Name
Parathion PESTANAL®, analytical standard
Product Number
45607
Product Brand
SIAL
CAS Number
56-38-2
Molecular Weight
291.26
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow to Brown-Yellow
APPEARANCE (FORM)Liquid
PURITY (HPLC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.5340 - 1.5380
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS