Product Name
Styrene analytical standard
Product Number
45993
Product Brand
SIAL
CAS Number
100-42-5
Molecular Weight
104.15
Test
Specification
APPEARANCE (COLOR)Colorless to Light Green-Yellow to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.5445 - 1.5485
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS