Product Name
9-Fluorenylmethyl carbazate for HPLC derivatization, LiChropur, ≥99.0%
Product Number
46917
Product Brand
SIAL
CAS Number
35661-51-9
Molecular Weight
254.28
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 99.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD54 MONTHS