Product Name
[(3R)-3-Hydroxydodecanoyl]-L-carnitine analytical standard
Product Number
49853
Product Brand
SIAL
CAS Number
1469900-99-9
Molecular Weight
359.50
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Beige to Beige
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
PURITY (HPLC AREA %)≥ 95.0 %
IDENTIFICATION (LC-MS)Identity confirmed
SPECIFIC ROTATION (20/D)-20 ± 3 Degrees
CONCENTRATIONc=0.1 in methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS