Product Name
QC-Standard Solution for EN ISO 9377-2 analytical standard
Product Number
51706
Product Brand
SIAL
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
REMARKS ON GCCONTAINS MINERAL OIL TYPE A AND B ~5 MG/ML EACH(BAM REFERENCE MATERIAL 5002 AND 5003) IN N-HEPTANE
DENSITY D20/40.686 - 0.690
EXPIRY PERIOD36 MONTHS