Product Name
Heptane analytical standard
Product Number
51730
Product Brand
SIAL
CAS Number
142-82-5
Molecular Weight
100.20
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.8 %
REFRACTIVE INDEX N20/D1.387 - 1.389
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS