Product Name
[(3R)-3-Hydroxyoctadecanoyl]-L-carnitine analytical standard
Product Number
52413
Product Brand
SIAL
CAS Number
1469901-05-0
Molecular Weight
443.66
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 94.0 %
SPECIFIC ROTATION (20/D)-19 ± 2 Degrees
CONCENTRATIONc=0.1 in methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS