Product Name
Hexane analytical standard
Product Number
52750
Product Brand
SIAL
CAS Number
110-54-3
Molecular Weight
86.18
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.7 %
REFRACTIVE INDEX N20/D1.374 - 1.376
EXPIRY PERIOD36 MONTHS