Product Name
Propionyl-L-carnitine-(N-methyl-d3) analytical standard
Product Number
52941
Product Brand
SIAL
CAS Number
203806-01-3
Molecular Weight
220.28
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
IDENTIFICATION (LC-MS)Degree of Labelling: ≥ 99.0 %
PURITY (TLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-22.0 ± 2.0 Degrees
CONCENTRATIONC=0.1 in Water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS