Product Name
[(3R)-3-Hydroxyhexanoyl]-L-carnitine analytical standard
Product Number
54253
Product Brand
SIAL
CAS Number
1469900-93-3
Molecular Weight
275.34
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-30.0 - -26.0 Degrees
CONCENTRATIONC=0.1 in Water
WATERWater <= 15 % (Calculated from Elemental Analysis)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS