Product Name
N-Stearoylglycine analytical standard
Product Number
54364
Product Brand
SIAL
CAS Number
6333-54-6
Molecular Weight
341.53
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS