Product Name
Silica gel high-purity grade, pore size 60 Å, 220-440 mesh particle size, 35-75 μm particle size, for flash chromatography
Product Number
60738
Product Brand
SIAL
CAS Number
112926-00-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
PARTICLE SIZECORRESPONDS TO REQUIREMENTS
LOSS ON DRYING≤ 5.0 %
RECOMMENDED RETEST PERIOD36 MONTHS