Product Name
Isovaleraldehyde analytical standard
Product Number
61848
Product Brand
SIAL
CAS Number
590-86-3
Molecular Weight
86.13
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
GC MINOR COMPONENT 1 (AREA %)0.0 - 3.0 %
MINOR COMPONENT 1ISOVALERIC ACID
REFRACTIVE INDEX N20/D1.386 - 1.390
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS