Product Name
m-Cresol analytical standard
Product Number
65996
Product Brand
SIAL
CAS Number
108-39-4
Molecular Weight
108.14
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.0 %
REFRACTIVE INDEX N20/D1.541 - 1.543
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS