Product Name
2-Methyl-2-butene analytical standard
Product Number
66050
Product Brand
SIAL
CAS Number
513-35-9
Molecular Weight
70.13
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.0 %
REMARKS ON GC2-Methyl-1-butene: Report Result
REFRACTIVE INDEX N20/D1.386 - 1.388
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS