Product Name
Isovitexin analytical standard
Product Number
67135
Product Brand
SIAL
CAS Number
38953-85-4
Molecular Weight
432.38
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
WATER≤ 12.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS