Product Name
2-Ethylhexanal analytical standard
Product Number
68155
Product Brand
SIAL
CAS Number
123-05-7
Molecular Weight
128.21
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 95.0 %
REFRACTIVE INDEX N20/D1.412 - 1.420
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS