Product Name
Alkane standard mixture for performance tests of GC-systems analytical standard, C10 - C40 (all even), 50 mg/l each
Product Number
68281
Product Brand
SIAL
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
REMARKS ON GCCONTAINS ALL EVEN N-ALKANES FROM C10 TO C40, 50 MG/L EACH
DENSITY D20/40.682 - 0.684
REFRACTIVE INDEX N20/D1.387 - 1.389
EXPIRY PERIOD36 MONTHS