Product Name
Sunset Yellow FCF analytical standard
Product Number
68775
Product Brand
SIAL
CAS Number
2783-94-0
Molecular Weight
452.37
Test
Specification
APPEARANCE (COLOR)Orange to Very Dark Orange and Red to Very Dark Red
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
ASSAY (ICP)Sodium (Theory): 10.16 %
RECOMMENDED RETEST PERIOD36 MONTHS