Product Name
2-Naphthol fluorescence indicator, ≥99.0%
Product Number
70450
Product Brand
SIAL
CAS Number
135-19-3
Molecular Weight
144.17
Test
Specification
APPEARANCE (COLOR)Off White to Beige and Faint Brown to Light Brown
APPEARANCE (FORM)Powder or Flakes
PURITY (GC AREA %)≥ 99.0 %
MELTING POINT121 - 122 C
SOLUBILITY (COLOR)Colorless to Light Yellow and Colorless to Light Brown-Yellow
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)100 mg/ml in methanol
RESIDUE ON IGNITION≤ 0.05 %
CARBON CONTENT83.31 % (theory)
HYDROGEN CONTENT5.59 % (theory)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM419 ± 5 nm
REMARKS ON LUMINESCENCEExc. 281 nm in 0.1 M NaOH