Product Name
Sodium selenite anhydrous, ≥90.0% (RT)
Product Number
71950
Product Brand
SIAL
CAS Number
10102-18-8
Molecular Weight
172.94
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder
REDOX TITRATION90.0 - 110.0 %
REDOXTITRATION (METHOD)IODOMETRIC
LOSS ON DRYING≤ 0.5%, 200C
RECOMMENDED RETEST PERIOD72 MONTHS