Product Name
trans-2-Dodecenoyl-L-carnitine analytical standard
Product Number
74023
Product Brand
SIAL
CAS Number
1631992-06-7
Molecular Weight
341.49
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 93.0 %
SPECIFIC ROTATION (20/D)-15 ± 2 Degrees
CONCENTRATIONc=0.1 in methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS