Product Name
Octane analytical standard
Product Number
74820
Product Brand
SIAL
CAS Number
111-65-9
Molecular Weight
114.23
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.7 %
REFRACTIVE INDEX N20/D1.396 - 1.398
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS