Product Name
1-Hexanethiol analytical standard
Product Number
75018
Product Brand
SIAL
CAS Number
111-31-9
Molecular Weight
118.24
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.448 - 1.452
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS