Product Name
3-Methoxytyramine hydrochloride analytical standard
Product Number
75024
Product Brand
SIAL
CAS Number
1477-68-5
Molecular Weight
203.67
Test
Specification
APPEARANCE (COLOR)White to Light Brown
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 96.0 %
CARBON CONTENT53.08% (Theory)
HYDROGEN CONTENT6.93% (Theory)
NITROGEN CONTENT6.88% (Theory)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
REMARKS≤ 1.0% Water (calculated from elemental analysis)
EXPIRY PERIOD36 MONTHS