Product Name
Oxalic acid puriss. p.a., anhydrous, ≥99.0% (RT)
Product Number
75688
Product Brand
SIAL
CAS Number
144-62-7
Molecular Weight
90.03
Test
Specification
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 50 mg/kg
MAGNESIUM (ICP)≤ 5 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
SODIUM (ICP)≤ 50 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 50 mg/kg
CHLORIDE (CL)≤ 50 mg/kg
TOTAL NITROGEN≤ 10 mg/kg
RECOMMENDED RETEST PERIOD54 MONTHS