Product Name
Vincamine analytical standard
Product Number
75778
Product Brand
SIAL
CAS Number
1617-90-9
Molecular Weight
354.44
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
TITRATION (NT) HCLO4 0.1M97.0 - 103.0 %
SPECIFIC ROTATION (20/D)43.0 ± 2.0 Degrees
CONCENTRATIONc=1 in pyridine
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS