Product Name
Piperidine solution suitable for peptide synthesis, 20% in DMF
Product Number
80645
Product Brand
SIAL
CAS Number
110-89-4
Molecular Weight
85.15
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow and Colorless to Very Faint Green-Yellow
APPEARANCE (FORM)Liquid
TITRATION (T) HCL 1M18.0 - 22.0 %
DENSITY D20/40.928 - 0.932
REFRACTIVE INDEX N20/D1.430 - 1.440
WATER (COULOMETR.)≤ 0.1 %
RECOMMENDED RETEST PERIOD36 MONTHS