Product Name
Polyvinylpyrrolidone K 30
Product Number
81420
Product Brand
SIAL
CAS Number
9003-39-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)Colorless to Almost Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.5G IN 10ML WATER
RESIDUE ON IGNITION≤ 0.05 %
CARBON CONTENT64.84 % (THEORY)
HYDROGEN CONTENT8.16 % (THEORY)
NITROGEN CONTENT12.60 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE