Product Name
Dimethyl terephthalate purum, ≥99.0% (GC)
Product Number
86440
Product Brand
SIAL
CAS Number
120-61-6
Molecular Weight
194.18
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)powder with chunks or briquettes
PURITY (GC AREA %)≥ 99.0 %
MELTING POINT140 - 142 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE