Product Name
p-Terphenyl suitable for scintillation, ≥98.5% (HPLC)
Product Number
86470
Product Brand
SIAL
CAS Number
92-94-4
Molecular Weight
230.30
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.5 %
MELTING POINT212 - 214 C
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.1G IN 10ML CHLOROFORM
INFRARED SPECTRUMCONFORMS TO STRUCTURE
WAVELENGTH (1) (UV)276 - 280 nm
ASSAY (1) (UV)≥ 98.5 %
SOLVENT (UV)not specified
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM343 ± 5 nm
RECOMMENDED RETEST PERIOD54 MONTHS