Product Name
Tetrabutylammonium hydroxide solution ~40% in water, suitable for ion chromatography
Product Number
86854
Product Brand
SIAL
CAS Number
2052-49-5
Molecular Weight
259.47
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow
APPEARANCE (FORM)Liquid
TITRATION (T) HCL 1M37.0 - 43.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
POTASSIUM (ICP)≤ 100 mg/kg
SODIUM (ICP)≤ 100 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 100 mg/kg
BROMIDE (BR)≤ 100 mg/kg
RECOMMENDED RETEST PERIOD42 MONTHS