Product Name
Tetracontane analytical standard
Product Number
87086
Product Brand
SIAL
CAS Number
4181-95-7
Molecular Weight
563.08
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals or Flakes
PURITY (GC AREA %)≥ 98.0 %
MELTING POINT80 - 83 C
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS