Product Name
3-Buten-2-ol analytical standard
Product Number
88497
Product Brand
SIAL
CAS Number
598-32-3
Molecular Weight
72.11
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS