Product Name
p-Toluidine for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Product Number
89630
Product Brand
SIAL
CAS Number
106-49-0
Molecular Weight
107.15
Test
Specification
APPEARANCE (COLOR)White to Beige
APPEARANCE (FORM)Crystals or Crystalline Chunk(s) or Chunk(s) or Solid
TITRATION (NT) HCLO4 0.1M99.0 - 101.0 %
PURITY (GC AREA %)≥ 99.0 %
MELTING POINT42 - 45 C
SOLUBILITY (COLOR)Colorless to Faint Brown and Colorless to Faint Brown-Yellow
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1G IN 10ML ETHANOL, HOT
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD24 MONTHS