Product Name
L-Carnitine-(trimethyl-13C3) inner salt hydrate analytical standard
Product Number
89927
Product Brand
SIAL
CAS Number
Molecular Weight
164.18 (anhydrous basis)
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
IDENTIFICATION (LC-MS)Degree of Labelling: ≥ 99.0 %
PURITY (TLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-35.0 - -29.0 Degrees
CONCENTRATIONc=0.1 in water
WATER≤ 15.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS