Product Name
3-Hydroxyisovaleryl-L-carnitine analytical standard
Product Number
91298
Product Brand
SIAL
CAS Number
99159-87-2
Molecular Weight
261.31
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Solid or Crystals
PURITY (HPLC AREA %)≥ 90.0 %
SPECIFIC ROTATION (20/D)-17.0 ± 2.0 Degrees
CONCENTRATIONc=0.1 in water
REM. ON ELEMENTAL ANALYSISwater calculated ≤ 15 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE