Product Name
cis-3-Hexen-1-ol analytical standard
Product Number
91316
Product Brand
SIAL
CAS Number
928-96-1
Molecular Weight
100.16
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.0 %
REFRACTIVE INDEX N20/D1.439 - 1.441
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS