Product Name
Hexanoyl-L-carnitine analytical standard
Product Number
91388
Product Brand
SIAL
CAS Number
22671-29-0
Molecular Weight
259.34
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-23.0 - -19.0 Degrees
CONCENTRATIONC=0.1 IN WATER
WATER≤ 10.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS